Order Entry
Norway
ContactUsLinkComponent
USB User-ID stick (with User-ID licence): Oven-linked authorisation licence (User-ID-programme) on Memory-stick, prevents undesired manipulation by unauthorised third parties. When reordering please specify serial number (only for units with TwinDISP 1 * 1 items
  MEMMB33170UF260TS
 :  MEMMERT
USB User-ID stick (with User-ID licence): Oven-linked authorisation licence (User-ID-programme) on Memory-stick, prevents undesired manipulation by unauthorised third parties. When reordering please specify serial number (only for units with TwinDISP 1 * 1 items
  MEMMB33170UF260TS
 :  MEMMERT
Frequently Bought Together