Flexible Pt100 temperature sensor, positioned flexibly in chamber or load, for local temperature measurement (up to 3 additional sen sors are possible). The measured temperature can, if required, be indicated on the display, recorded in the integral 1 * 1 items
Catalog # MEMMUF750TSH8
Supplier: MEMMERT
Frequently Bought Together