Phloretin 98%
Glass bottle, 500 mg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:60-82-2
- Formula:C₁₅H₁₄O₅
- MDL Number:MFCD00002288
- Molecular Weight:274.28 g/mol
- Melting Pt:260…262 °C
- Storage Temperature:Freezer
- EINECS:200-488-7
Documents
ACRO30765
SDS
Specifications
Specification Test Results
- Appearance:White to beige Powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
Specifications
- Pack type:Glass bottle
- Pk:500 mg



