Bly(II)nitrat 99% for analysis
Plastic bottle, 250 g
Supplier: Thermo Fisher Scientific
Bly(II)nitrat 99% for analysis
Catalog # ACRO315242500
Supplier: Thermo Fisher Scientific
CAS Number: 10099-74-8
Jump to:
Product Details & Documents
Synonyms: Lead dinitrate
- CAS Number:10099-74-8
- Formula:Pb(NO₃)₂
- MDL Number:MFCD00011153
- Molecular Weight:331.21 g/mol
- Melting Pt:470 °C (dec.)
- Density:4,53 g/cm³
- Storage Temperature:Romtemperatur
- UN:1469
- ADR:5.1,II
Documents
ACRO31524
SDS
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Titration Complexometric:≥98.5 %
- Chloride (Cl):≤0.005 %
- Copper (Cu):≤0.001 %
- Iron (Fe):≤0.001 %
- Insoluble matter:≤0.005 % (10 % in dilute HNO3)
Specifications
- Pakketype:Plastic bottle
- Fp:250 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



