Lead(II) nitrate 99% for analysis
Plastic bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
Lead(II) nitrate 99% for analysis
Catalog # ACRO315240025
Supplier: Thermo Fisher Scientific
CAS Number: 10099-74-8
Jump to:
Product Details & Documents
Synonyms: Lead dinitrate
- CAS Number:10099-74-8
- Formula:Pb(NO₃)₂
- MDL Number:MFCD00011153
- Molecular Weight:331.21 g/mol
- Melting Pt:470 °C (dec.)
- Density:4.53 g/cm³
- Storage Temperature:Ambient
- UN:1469
- ADR:5.1,II
Documents
ACRO31524
SDS
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Titration Complexometric:≥98.5 %
- Chloride (Cl):≤0.005 %
- Copper (Cu):≤0.001 %
- Iron (Fe):≤0.001 %
- Insoluble matter:≤0.005 % (10 % in dilute HNO3)
Specifications
- Pack type:Plastic bottle
- Pk:2,5 kg



