4'-Bromoacetophenone 98%
Glass bottle, 500 g
Supplier: Thermo Fisher Scientific
4'-Bromoacetophenone 98%
Catalog # ACRO106595000
Supplier: Thermo Fisher Scientific
CAS Number: 99-90-1
Gå til:
Product Details & Documents
Synonymer: p-Bromoacetophenone, 1-(4-Bromophenyl)ethanone
- CAS Number:99-90-1
- Formel:C₈H₇BrO
- MDL-nummer:MFCD00000105
- Molecular Weight:199.05 g/mol
- Merck-indeksen:13,01387
- Kokende Pt:255 °C (760 mmHg)
- Molten Pt:48…52 °C
- Tetthet:1,736 g/cm³ (20 °C)
- Flash Pt:>100 °C
- Lagringstemperatur:Ambient
- EINECS:202-799-3
- ADR:6.1,III
Documents
ACRO10659
SDS
Spesifikasjoner
Spesifikasjonstestresultater
- Appearance:White to light yellow Crystals or flakes
- Infrared spectrum:Conforms
- Melting point:48 to 52 °C
- GC:≥97.5 %
Spesifikasjoner
- Pack type:Glass bottle
- Pk:500 g
Spesifikasjoner
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



