kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
PAL-COFFEE Brix 0.00 to 25.00% , +/- 0,10%, +/- 1C 1 * 1 items
ATAG4523
:
Atago
PAL-COFFEE Brix 0.00 to 25.00% , +/- 0,10%, +/- 1C 1 * 1 items
ATAG4523
:
Atago