Order Entry
Norway
Orders LinkContactUsLinkComponent
 

Single - component oxygenates solutions

tert-Amyl methyl ether (TAME), with 2000 µg/ml in purge and trap grade methanol, US EPA 502.2, 8021, 8206B, 1 ml/ampoule, 1 Ampoul
  REST30629
 :   Restek
Single - component oxygenates solutions
  REST30629
 :  Restek
 :  
Single - component oxygenates solutions
Single - component oxygenates solutions

tert-Amyl methyl ether (TAME), with 2000 µg/ml in purge and trap grade methanol, US EPA 502.2, 8021, 8206B, 1 ml/ampoule, 1 Ampoul

 :  
 
kr 935,00
   
1 * 1 Each
This product has been blocked by your organization. Please contact your purchasing department for more information
 

Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.

 

 

  • Pk:
    1 Ampoul
  • Standards:
    US EPA 502.2, 8021, 8206B
  • Description:
    tert-Amyl methyl ether (TAME), with 2000 µg/ml in purge and trap grade methanol
  • Pk Info:
    1 ml/ampoule
  • Cat. No.:
    REST30629
  • Formula:
    C₆H₁₄O
  • Description:
    tert-Amyl methyl ether (TAME)
  • CAS No.:
    994-05-8

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.