Single - component oxygenates solutions
tert-Butanol-d9, with 20000 µg/ml in purge and trap grade methanol, US EPA 502.2, 8021, 8206B, 1 ml/ampoule, 1 Ampoul
Catalog # REST30618
Supplier: Restek
Jump to:
Product Details & Documents
Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Specifications
Specifications
- Pk:1 Ampoul
- Standards:US EPA 502.2, 8021, 8206B
- Description:tert-Butanol-d9, with 20000 µg/ml in purge and trap grade methanol
- Pk Info:1 ml/ampoule
- Cat. No.:REST30618
- Formula:(CD₃)₃COH
- Description:tert-Butanol-d9
- CAS No.:25725-11-5



