Automation Tips, GripTips
Product Details & Documents
GRIPTIPS are suitable for use with the benchtop pipetting platforms MINI 96, VIAFLO 96, VIAFLO 384 And ASSIST PLUS.
- Tips perfectly match the multi-lobe tip fitting, snapping firmly on during loading
- Perfect seal on every tip, preventing them from loosening, leaking or completely falling off
- All tips are precisely aligned horizontally, enabling accurate touch-offs, even when pipetting with 384 tips
The tips undergo additional quality control procedures – such as a secondary straightness test – to guarantee perfect targeting of wells for automatic pipetting or simultaneous pipetting with up to 384 tips. The tip racks are also reinforced to withstand the forces required to load 384 tips.
Packaging: GRIPTIP Packaging Options
1) Tip Racks with single inserts in a 96-tip configuration – Reinforced Tip Racks for 300 and 1,250 µl straightness tested GRIPTIPS for use with 96-channel pipetting heads and automated multichannel pipettes on the ASSIST/ASSIST PLUS.
2) XYZ Tip Racks – Extra sturdy and in an SBS format to withstand the loading forces from automated multichannel pipettes (ASSIST/ASSIST PLUS), and 96 and 384channel pipetting heads (12,5 to 125 µl tips). GRIPTIPS are ionized to dissipate existing static, and then loaded into carbon-filled, antistatic XYZ boxes.
3) GREEN CHOICE refill system – an environmentally friendly refill system allows you to reuse your existing Tip Racks, reducing plastic waste by 60 % (300 and 1250 µl tips only).
Documents
Certificates
Search by Lot NumberSpecifications
- Packed:5 XYZ racks of 384 tips
- Sterile:-
- Volume:12,5 µl
- Seq:19
- Tip style:GripTips, long
- Filter:-