Order Entry
Norway
ContactUsLinkComponent
Cleanroom tape, model 1112, white, width 4 (102mm), length 36yds (33m) Polyethylene film with medium-tack no-residue acrylic adhesive Double-bagged individually 1 * 1 Roll
Catalog # UTAP1112WH400-P3T
Supplier:  ULTRATAPE
Cleanroom tape, model 1112, white, width 4 (102mm), length 36yds (33m) Polyethylene film with medium-tack no-residue acrylic adhesive Double-bagged individually 1 * 1 Roll
Catalog # UTAP1112WH400-P3T
Supplier:  ULTRATAPE
Frequently Bought Together