0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Rat MMP 8 ELISA Kit for Serum, Plasma, Cell Culture Supernatant. 1 * 96 items
Katalog # BIRBORB79130-96
Supplier:
Biorbyt
Expand image
Rat MMP 8 ELISA Kit for Serum, Plasma, Cell Culture Supernatant. 1 * 96 items
Katalog # BIRBORB79130-96
Supplier:
Biorbyt
Related Information
Documentation
Datasheet
orb79130
Documentation