Order Entry
Norway
Orders LinkContactUsLinkComponent
LC3 (from<i> E. coli</i>)

LC3 (from E. coli)

5 µG
Catalog # BIRBORB81209-5
Marketsource
Supplier:   Biorbyt
LC3 (from<i> E. coli</i>)
Catalog # BIRBORB81209-5
Supplier:  Biorbyt
Marketsource
CAS Number:  
LC3 (from E. coli)

5 µG

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 Each
Product Details & Documents

Documents

Specifications

Specifications

  • Pk:
    5 µG
  • Cat. No.:
    BIRBORB81209-5
  • Protein/peptide name:
    LC3
  • Storage conditions:
    MAP1LC3A Human although stable at 4 °C for week, should be stored desiccated below −18 °C. Please prevent freeze/thaw cycles.
  • Formulation:
    1mg/ml solution containing 20 mM Tris pH-8, 1mM DTT, 0.1M NaCl 10% glycerol.
  • Protein synonyms:
    ATG8F
  • Purity:
    Greater than 95.0% as determined by SDS-PAGE.
  • Sequence:
    MPSDRPFKQR RSFADRCKEV QQIRDQHPSK IPVIIERYKG EKQLPVLDKT KFLVPDHVNM SELVKIIRRR LQLNPTQAFF LLVNQHSMVSVSTPIADIYE QEKDEDGFLY MVYASQETFG LEHHHHHH.
  • Tested applications:
    SDS-PAGE
  • Source:
    E. coli

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.