Order Entry
Norway
Orders LinkContactUsLinkComponent
 

LC3 (from E. coli)

20 µG
Catalog # BIRBORB81209-20
Marketsource
Supplier:   Biorbyt
LC3 (from<i> E. coli</i>)
Catalog # BIRBORB81209-20
Supplier:  Biorbyt
Marketsource
CAS Number:  
LC3 (from E. coli)

20 µG

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 Each
Product Details & Documents

 

Specifications

Specifications

  • Pk:
    20 µG
  • Cat. No.:
    BIRBORB81209-20
  • Protein/peptide name:
    LC3
  • Storage conditions:
    MAP1LC3A Human although stable at 4 °C for week, should be stored desiccated below −18 °C. Please prevent freeze/thaw cycles.
  • Formulation:
    1mg/ml solution containing 20 mM Tris pH-8, 1mM DTT, 0.1M NaCl 10% glycerol.
  • Protein synonyms:
    ATG8F
  • Purity:
    Greater than 95.0% as determined by SDS-PAGE.
  • Sequence:
    MPSDRPFKQR RSFADRCKEV QQIRDQHPSK IPVIIERYKG EKQLPVLDKT KFLVPDHVNM SELVKIIRRR LQLNPTQAFF LLVNQHSMVSVSTPIADIYE QEKDEDGFLY MVYASQETFG LEHHHHHH.
  • Tested applications:
    SDS-PAGE
  • Source:
    E. coli

Specifications