Order Entry
Norway
ContactUsLinkComponent
Recombinant Full-Length H3N2 A/Wisconsin/67/05 is glycosylated with N-linked sugars, produced using baculovirus vectors in insect ce lls and its Mw is 70,000 dalton. 1 * 10 µG
Catalog # BIRBORB81784-10
Supplier:  Biorbyt
Recombinant Full-Length H3N2 A/Wisconsin/67/05 is glycosylated with N-linked sugars, produced using baculovirus vectors in insect ce lls and its Mw is 70,000 dalton. 1 * 10 µG
Catalog # BIRBORB81784-10
Supplier:  Biorbyt
Related Information