Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
LDPE tubefoil 600mm x 0 15mm Roll 200 meters 600 x 0.15 mm, 1 * 1 Roll
Catalog # KWPA122829
Supplier:
KWP KUNSTSTOFF WERK PLUR
Expand image
LDPE tubefoil 600mm x 0 15mm Roll 200 meters 600 x 0.15 mm, 1 * 1 Roll
Catalog # KWPA122829
Supplier:
KWP KUNSTSTOFF WERK PLUR