Order Entry
Norway
ContactUsLinkComponent
LDPE tubefoil flat 600x100x0 1 mmm antistatic, transparent,Cleanroom class ISO 5 (Roll = 250m) 1 * 1 Roll
Catalog # KWPA121554
LDPE tubefoil flat 600x100x0 1 mmm antistatic, transparent,Cleanroom class ISO 5 (Roll = 250m) 1 * 1 Roll
Catalog # KWPA121554