Order Entry
Norway
Orders LinkContactUsLinkComponent
Goniometer, digital, used for experiments on prism or grating deviation, 20 seconds of arc, consist of autocollimating telescope, ad justable focus by knurled screw, focus length 162mm, sliding eyepiece, collimator manual setting °. 1 * 1 items
Goniometer, digital, used for experiments on prism or grating deviation, 20 seconds of arc, consist of autocollimating telescope, ad justable focus by knurled screw, focus length 162mm, sliding eyepiece, collimator manual setting °. 1 * 1 items
Catalog # DIDAPOD068090
Supplier:   DIDALAB
Goniometer, digital, used for experiments on prism or grating deviation, 20 seconds of arc, consist of autocollimating telescope, ad justable focus by knurled screw, focus length 162mm, sliding eyepiece, collimator manual setting °. 1 * 1 items
Catalog # DIDAPOD068090
Supplier:  DIDALAB
Goniometer, digital, used for experiments on prism or grating deviation, 20 seconds of arc, consist of autocollimating telescope, ad justable focus by knurled screw, focus length 162mm, sliding eyepiece, collimator manual setting °. 1 * 1 items
Price
Inquire For Price
Log in for availability
1 * 1 Items