Order Entry
Norway
Orders LinkContactUsLinkComponent
Goniometer, basic, specially adapted to high schools experiments (prism and grating deviations, it has a telescope and a collimator (focus lengths 178mm), this device is made up of a diameter 127mm graduated plate. 1 * 1 items
Goniometer, basic, specially adapted to high schools experiments (prism and grating deviations, it has a telescope and a collimator (focus lengths 178mm), this device is made up of a diameter 127mm graduated plate. 1 * 1 items
Catalog # DIDAPOD068010
Supplier:   DIDALAB
Goniometer, basic, specially adapted to high schools experiments (prism and grating deviations, it has a telescope and a collimator (focus lengths 178mm), this device is made up of a diameter 127mm graduated plate. 1 * 1 items
Catalog # DIDAPOD068010
Supplier:  DIDALAB
Goniometer, basic, specially adapted to high schools experiments (prism and grating deviations, it has a telescope and a collimator (focus lengths 178mm), this device is made up of a diameter 127mm graduated plate. 1 * 1 items
Price
kr12,700.00
Log in for availability
1 * 1 Items