Order Entry
Norway
Orders LinkContactUsLinkComponent
Labels, pipe marking
Labels, pipe marking
Pipe marking labels with GHS04 symbol, Polyester, Black on yellow, 250×26 mm, 4 labels/card
Catalog # BRDYN002355
Labels, pipe marking
Catalog # BRDYN002355
Supplier:  
Labels, pipe marking
Labels, pipe marking
Pipe marking labels with GHS04 symbol, Polyester, Black on yellow, 250×26 mm, 4 labels/card
Discontinued
Product Details & Documents
These pipe marking labels are made of high quality polyester and meet with GHS (Global Harmonised System) regulations for chemical substances and mixtures.
  • Promotes safety and simplifies maintenance
  • Arrows are pre-cut, and hence does not requires scissors additionally
  • Resistant to temperatures from –40 to +120 °C
  • Resistant to ultraviolet rays and conditions
The markers have a frame that contrasts with the color of the text to increase visibility.
Specifications
  • Colour:
    Black on yellow
  • Description:
    Pipe marking labels with GHS04 symbol
  • Dimensions:
    250×26 mm
  • Material:
    Polyester
  • Seq:
    07
  • Packed:
    4 labels/card
  • Cat. No.:
    BRDYN002355