Order Entry
Norway
ContactUsLinkComponent
Calibration gas 25 h2s,100co,1.25%ch4,18%o2,n2 34l 4g18x125w25h100-34 1 * 1 items
Catalog # BWTE4G18X125W25H10
Supplier:  BW TECHNOLOGIES
Calibration gas 25 h2s,100co,1.25%ch4,18%o2,n2 34l 4g18x125w25h100-34 1 * 1 items
Catalog # BWTE4G18X125W25H10
Supplier:  BW TECHNOLOGIES