Inserts
E 364, insert 1/2, with 36 compartments, for 36 wide neck vaccums, PG 8535/36, PG 8581, PG 8582, PG 8582 CD, PG 8591, PG 8592, 215×445×77 mm
Catalog # MIEL3809640
Supplier: MIELE
Jump to:
Product Details & Documents
Specifications
- Description:E 364, insert 1/2, with 36 compartments, for 36 wide neck vaccums
- Seq:10
- W×D×H:215×445×77 mm
- For:PG 8535/36, PG 8581, PG 8582, PG 8582 CD, PG 8591, PG 8592