Order Entry
Norway
ContactUsLinkComponent
Laboratory Chair Neon, Grey, Weight Regulation, Seat Depth Adjustment, Backrest Height Adjustment, Seat Height Adjustment With A Ga sspring From 450-620 Mm, 5-Star Base Made Of Aluminium, Powder Coated Black, With Stop-And-Go-Castors, With Foot Ring 1 * 1 items
Catalog # BIMO571MG113278807
Supplier:  bimos
Laboratory Chair Neon, Grey, Weight Regulation, Seat Depth Adjustment, Backrest Height Adjustment, Seat Height Adjustment With A Ga sspring From 450-620 Mm, 5-Star Base Made Of Aluminium, Powder Coated Black, With Stop-And-Go-Castors, With Foot Ring 1 * 1 items
Catalog # BIMO571MG113278807
Supplier:  bimos