Order Entry
Norway
ContactUsLinkComponent
1 perforated metal plate insert for pull out sump, additional second level drawer, perforated plate: stainless steel 1.4301, second level drawer: plastic laminated steel sheet ral 7035 1 * 1 items
Catalog # MSGA30460
Supplier:  asecos
1 perforated metal plate insert for pull out sump, additional second level drawer, perforated plate: stainless steel 1.4301, second level drawer: plastic laminated steel sheet ral 7035 1 * 1 items
Catalog # MSGA30460
Supplier:  asecos