Colony counters, ProtoCOL 3
Automated colony counter with zone sizing, validation plates, and software, 3 plus
SYNBPROC3PLUS
ProtoCOL 3 is designed for colony counting, zone measuring (inhibition and AST), membranes, Petri-film™ and other applications including spiral plates, OPKA, SBA, Multi-sector, Multi-well, SRD and Ames.
- High throughput capability suitable for production and large scale testing environments
- Extensive report generation to Excel or a LIMS system
- Separation of touching colonies, exclusion areas, colour mode, shape mode, size mode
- High resolution colour images ensure that the smallest colony can be seen and counted with zone measurements
- Mounted touch screen processor, which enables ease of control while the easy to use software produces highly accurate results
Designed for use across a wide range of applications, ProtoCOL 3 will count colonies as small as 43 µm (0,043 mm) or measure zones accurately to 0,1 mm.
: GLP/GMP compliant data.
- Model:3 plus
- Description:Automated colony counter with zone sizing, validation plates, and software
- Cat. No.:SYNBPROC3PLUS
- Dish size:150 mm
- W×D×H:400×500×450 mm
- Illumination:Two sliding doors to prevent excessive ambient light
Unique three channels: Red, green, blue
LED lighting (patent pending)
Multi-array computer controlled LED
Lower lighting with upper reflective lighting for all applications - Camera:Scientific grade CCD
- Weight:20 kg
- Resolution:1,4 MP
Accessories
- Description
- Description
- Description
- SYNBPROC3SECTORMODMulti-sector plates software module for counting on multi-sector plates, eg, air sample1 * 1 Items - kr 15 800,00
- Description
- SYNBPROC3-AMES-MODAmes software module for the bacterial reverse mutation test1 * 1 Items - kr 15 800,00
- Description
- SYNBPROC3SPIRALMODCounting software module for spiral plates, includes validation plate1 * 1 Items - kr 15 500,00
- Description
- SYNBPROC3-OPKA-MODOPKA software module for opsonophagocytic killing assay1 * 1 Items - kr 15 800,00



