kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
REFRACTIVE INDEX STD SERIE B RI=1,670 1 * 7,4 mL
CARG1812X1.670
:
CARGILLE
REFRACTIVE INDEX STD SERIE B RI=1,670 1 * 7,4 mL
CARG1812X1.670
:
CARGILLE