Semi Dry Blot Transfer buffer (10X)
Semi-dry Blot Transfer Buffer, 10X, 2 L
Supplier: Thermo Fisher Scientific
Semi Dry Blot Transfer buffer (10X)
Catalog # J63664.K3
Supplier: Thermo Fisher Scientific
CAS Number:
Gå til:
Product Details & Documents
Physical Form: Liquid
Documents
Spesifikasjoner
Spesifikasjoner
- Pk:2 L
- Description:Semi-dry Blot Transfer Buffer, 10X
- Cat. No.:J63664.K3
- Physical form:Liquid
Spesifikasjoner
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



