Potassium acetate 8 M in aqueous solution
Glass bottle, 250 mL
Supplier: Thermo Fisher Scientific
Potassium acetate 8 M in aqueous solution
Catalog # J63372.AK
Supplier: Thermo Fisher Scientific
CAS Number: 127-08-2
Jump to:
Product Details & Documents
CAS No.: 127-08-2 Molecular Formula: C2H3KO2 Formula Weight: 98.15 Physical Form: Liquid MDL No.: MFCD00012458
Synonyms: Acetic acid potassium salt
- CAS Number:127-08-2
- Formula:CH₃COOK
- MDL Number:MFCD00012458
- Molecular Weight:98.14 g/mol
- Storage Temperature:Ambient
- EINECS:204-822-2
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:250 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



