Potassium acetate 8 M in aqueous solution
Glass bottle, 100 mL
Supplier: Thermo Fisher Scientific
Potassium acetate 8 M in aqueous solution
Catalog # J63372.AE
Supplier: Thermo Fisher Scientific
CAS Number: 127-08-2
Jump to:
Product Details & Documents
CAS No.: 127-08-2 Molecular Formula: C2H3KO2 Formula Weight: 98.15 Physical Form: Liquid MDL No.: MFCD00012458
Synonyms: Acetic acid potassium salt
- CAS Number:127-08-2
- Formula:CH₃COOK
- MDL Number:MFCD00012458
- Molecular Weight:98.14 g/mol
- Storage Temperature:Ambient
- EINECS:204-822-2
Specifications
Specifications
- Pack type:Glass bottle
- Pk:100 mL



