Ginkgolide B
Glass bottle for solids, 10 mg
Supplier: Thermo Fisher Scientific
Gå til:
Product Details & Documents
CAS No.: 15291-77-7 Molecular Formula: C20H24O10 Formula Weight: 424.40 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White Solubility: Soluble in DMSO. Slightly soluble in water and ethanol Merck Reference: 14,4426 MDL No.: MFCD05662347
Platelet-Activating Factor (PAF) antagonist
- CAS Number:15291-77-7
- Formel:C₂₀H₂₄O₁₀
- MDL-nummer:MFCD05662347
- Molecular Weight:424.40 g/mol
- Merck-indeksen:14,4426
Documents
Spesifikasjoner
Spesifikasjoner
- Pack type:Glass bottle for solids
- Pk:10 mg
Spesifikasjoner
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



