HEPES, 1,0 M buffer solutions
HEPES buffer solution, 1 mol/L, 6,5, Glass bottle, 500 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 7365-45-9
MDL No.: MFCD00006158
Specifications
Specifications
- Concentration:1 mol/L
- Pack type:Glass bottle
- Pk:500 mL
- Description:HEPES buffer solution
- pH value:6,5
- Cat. No.:J61017.AP
- Physical form:Liquid
- MDL No.:MFCD00006158
- CAS No.:7365-45-9



