Shelving systems, solid, Super Erecta®
Product Details & Documents
Perfect for applications involving spillage. Provides barrier between floor and bottom shelf contents to deter contamination.
- Shelf design features a 3,2 mm raised 'ship’s edge' on all four sides to contain spills
- Louvered/embossed shelf allows air circulation
- Available in several styles and materials for a variety of applications
- Shelves are constructed with 18-gauge stainless steel (type 304) or galvanized materials
Standard stainless steel solid shelves (Type 304): with epoxy-coated cast corners address the majority of applications for solid, corrosion resistant shelves or work surfaces. Available in flat solid and louvered/embossed styles.
Galvanized shelves: with uncoated cast corners are ideal for applications requiring a solid shelving or work surface and minimum resistance to corrosion. Available in flat solid and louvered/embossed styles.
Premium Autoclave/Cart Washable Stainless Steel solid shelves (Type 304): feature an all-stainless construction to withstand high temperature, corrosive applications. Available in flat solid style.
Ordering information: Each individual shelf and post is sold separately. Accessories are also available.
Specifications
- Description:Super Erecta® autoclavable and cart-washable solid shelf, (width×length): 530×1524 mm, 2160NFS
- Material:Stainless steel 304
- Seq:58
- Weight:11,8 kg
Accessories
Product Information
- Catalog #AvailabilityPrice
- Label holders, for 75×32 mm labels
Product Information
- Catalog #ForAvailabilityPrice
- Mobile post, SiteSelect™, chrome plated, 1590 mm heightMobile post, SiteSelect™, chrome plated, 1590 mm height139-0268Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelvingLog in for availability1 * 1 Items - kr 585,00
Product Information
- Catalog #ForAvailabilityPrice
- Mobile post, SiteSelect™, chrome plated, 2454 mm heightMobile post, SiteSelect™, chrome plated, 2454 mm height139-0270Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelvingLog in for availability1 * 1 Items - kr 810,00
Product Information
- Catalog #ForAvailabilityPrice
- Mobile post, SiteSelect™, stainless steel, 2454 mm heightMobile post, SiteSelect™, stainless steel, 2454 mm height139-0277Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, Super Erecta Pro™ shelvingLog in for availability1 * 1 Items - kr 2 175,00
Product Information
- Catalog #AvailabilityPrice
- Slanted label holders, for 75×32 mm labelsSlanted label holders, for 75×32 mm labelsMETO9990SLLog in for availability1 * 1 Items - kr 97,00
Product Information
- Catalog #ForAvailabilityPrice
- Mobile post, SiteSelect™, stainless steel, 1590 mm heightMobile post, SiteSelect™, stainless steel, 1590 mm height139-0275Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, Super Erecta Pro™ shelvingLog in for availability1 * 1 Items - kr 1 625,00
Product Information
- Catalog #ForAvailabilityPrice
- Stationary post, SiteSelect™, chrome plated, 2200 mm heightStationary post, SiteSelect™, chrome plated, 2200 mm height139-0255Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelvingLog in for availability1 * 1 Items - kr 810,00
Product Information
- Catalog #ForAvailabilityPrice
- Stationary post, SiteSelect™, chrome plated, 875 mm heightStationary post, SiteSelect™, chrome plated, 875 mm height139-0251Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelvingLog in for availability1 * 1 Items - kr 495,00
Product Information
- Catalog #AvailabilityPrice
- Decorator shelf inlays, black/white, 610×610 mmDecorator shelf inlays, black/white, 610×610 mmMETO2424BWILog in for availability1 * 1 Items - kr 830,00
Product Information
- Catalog #AvailabilityPrice
- Plastic split sleeves, black
Product Information
- Catalog #AvailabilityPrice
- Clear label holders,1520 mm, for 1395×32 mm labelsClear label holders,1520 mm, for 1395×32 mm labelsMETO9990CL5Log in for availability1 * 1 Items - kr 490,00
Product Information
- Catalog #ForAvailabilityPrice
- Mobile post, SiteSelect™, chrome plated, 1880 mm heightMobile post, SiteSelect™, chrome plated, 1880 mm height139-0269Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelvingLog in for availability1 * 1 Items - kr 635,00
Product Information
- Catalog #AvailabilityPrice
- Aluminum split sleeves with stainless ringsAluminum split sleeves with stainless ringsMETO9986SLog in for availability1 * 1 Each - kr 1 400,00
Product Information
- Catalog #ForAvailabilityPrice
- Mobile post, SiteSelect™, stainless steel, 1880 mm heightMobile post, SiteSelect™, stainless steel, 1880 mm height139-0276Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, Super Erecta Pro™ shelvingLog in for availability1 * 1 Items - kr 1 775,00
Product Information
- Catalog #ForAvailabilityPrice
- Stationary post, SiteSelect™, stainless steel, 1590 mm heightStationary post, SiteSelect™, stainless steel, 1590 mm height139-0260Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, Stationary shelving, Super Erecta Pro™ shelving, Super Erecta Pro™ shelvingLog in for availability1 * 1 Items - kr 1 625,00
Product Information
- Catalog #ForAvailabilityPrice
- Stationary post, SiteSelect™, metroseal 3, 1590 mm heightStationary post, SiteSelect™, metroseal 3, 1590 mm heightMETO63PK3Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta Pro™ shelvingLog in for availability1 * 1 Items - kr 685,00
Product Information
- Catalog #AvailabilityPrice
- Enclosure panel, chrome, 313×1518 mm, for nominal post height 1600 mmEnclosure panel, chrome, 313×1518 mm, for nominal post height 1600 mmMETOEP36CLog in for availability1 * 1 Items - kr 1 550,00
Product Information
- Catalog #ForAvailabilityPrice
- Stationary post, SiteSelect™, chrome plated, 1590 mm heightStationary post, SiteSelect™, chrome plated, 1590 mm height139-0253Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelvingLog in for availability1 * 1 Items - kr 585,00
Product Information
- Catalog #ForAvailabilityPrice
- Stationary post, SiteSelect™, metroseal 3, 1895 mm heightStationary post, SiteSelect™, metroseal 3, 1895 mm heightMETO74PK3Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta Pro™ shelvingLog in for availability1 * 1 Items - kr 745,00
Product Information
- Catalog #AvailabilityPrice
- Post mounting shelf supports, width 530 mm, double, chrome platedPost mounting shelf supports, width 530 mm, double, chrome platedMETO2WS21CLog in for availability1 * 1 Items - kr 1 375,00
Product Information
- Catalog #ForAvailabilityPrice
- Stationary post, SiteSelect™, chrome plated, 1895 mm heightStationary post, SiteSelect™, chrome plated, 1895 mm height139-0254Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelvingLog in for availability1 * 1 Items - kr 635,00
Product Information
- Catalog #AvailabilityPrice
- Post mounting shelf supports, width 355 mm, double, chrome platedPost mounting shelf supports, width 355 mm, double, chrome platedMETO2WS14CLog in for availability1 * 1 Items - kr 1 325,00
Product Information
- Catalog #AvailabilityPrice
- Post mounting shelf supports, width 530 mm, single, chrome platedPost mounting shelf supports, width 530 mm, single, chrome platedMETO1WS21CLog in for availability1 * 1 Items - kr 930,00
Product Information
- Catalog #AvailabilityPrice
- Post mounting shelf supports, width 355 mm, single, chrome platedPost mounting shelf supports, width 355 mm, single, chrome platedMETO1WS14CLog in for availability1 * 1 Items - kr 895,00
Product Information
- Catalog #AvailabilityPrice
- Storage baskets, chrome plated, 190×440×225 mm, 5 kgStorage baskets, chrome plated, 190×440×225 mm, 5 kgMETOH212CLog in for availability1 * 1 Items - kr 1 250,00
Product Information
- Catalog #AvailabilityPrice
- Storage baskets, chrome plated, 127×345×180 mm, 4 kgStorage baskets, chrome plated, 127×345×180 mm, 4 kgMETOH209CLog in for availability1 * 1 Items - kr 885,00
Product Information
- Catalog #AvailabilityPrice
- Enclosure panel, chrome, 464×1791 mm, for nominal post height 1800 mmEnclosure panel, chrome, 464×1791 mm, for nominal post height 1800 mmMETOEP57CLog in for availability1 * 1 Items - kr 1 950,00
Product Information
- Catalog #AvailabilityPrice
- Additional tabs for rod, bag of 12
Product Information
- Catalog #MaterialAvailabilityPrice
- Clear shelf inlays for Super Erecta® shelving system, 914×355 mmClear shelf inlays for Super Erecta® shelving system, 914×355 mmMETO1436CI-4PlasticLog in for availability1 * 1 Each - kr 1 600,00
Product Information
- Catalog #AvailabilityPrice
- Hanger rail fits on posts along the width or length of the unit 457 mm, fits to size shelf width: 914 mm. Optional hooks can hang from rail at any point.Hanger rail fits on posts along the width or length of the unit 457 mm, fits to size shelf width: 914 mm. Optional hooks can hang from rail at any point.METOH118CLog in for availability1 * 1 Items - kr 470,00
Product Information
- Catalog #AvailabilityPrice
- Aluminum split sleeves with zinc rings perfect for high temperature or conductive applications ; come with zinc retainer rings to secure the shelf in place (one pack required/shelf)Aluminum split sleeves with zinc rings perfect for high temperature or conductive applications ; come with zinc retainer rings to secure the shelf in place (one pack required/shelf)METO9986ZLog in for availability1 * 1 Each - kr 235,00
Product Information
- Catalog #AvailabilityPrice
- Extension display hanger (152 mm): snaps on Super Erecta® shelf hanger rails and shelf frames, accepts standard carded merchandiseExtension display hanger (152 mm): snaps on Super Erecta® shelf hanger rails and shelf frames, accepts standard carded merchandiseMETOHHK6CLog in for availability1 * 1 Items - kr 250,00
Product Information
- Catalog #AvailabilityPrice
- Split sleeves, plastic, white
Product Information
- Catalog #AvailabilityPrice
- Slanted label holders, for 75×32 mm labelsSlanted label holders, for 75×32 mm labelsMETO9990CLLog in for availability1 * 1 Items - kr 97,00
Product Information
- Catalog #AvailabilityPrice
- Clear label holders, plastic, fits 914 mm shelfs, 725×32 mmClear label holders, plastic, fits 914 mm shelfs, 725×32 mmMETO9990CL3Log in for availability1 * 1 Items - kr 290,00
Product Information
- Catalog #MaterialAvailabilityPrice
- Clear shelf inlays for Super Erecta® shelving system, 914×530 mmClear shelf inlays for Super Erecta® shelving system, 914×530 mmMETO2136CI-4PlasticLog in for availability1 * 1 Each - kr 1 725,00
Product Information
- Catalog #ForAvailabilityPrice
- Swaged post, stainless steel with aluminium cap, 1600 mm dimensionSwaged post, stainless steel with aluminium cap, 1600 mm dimensionMETO63UPS-SWSuper Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta Pro™ shelvingLog in for availability1 * 1 Items - kr 1 775,00
Product Information
- Catalog #ForAvailabilityPrice
- Stationary post, SiteSelect™, chrome plated, 2454 mm heightStationary post, SiteSelect™, chrome plated, 2454 mm heightMETO96PDrop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelvingLog in for availability1 * 1 Items - kr 925,00
Product Information
- Catalog #ForAvailabilityPrice
- Mobile post, SiteSelect™, stainless steel, 699 mm heightMobile post, SiteSelect™, stainless steel, 699 mm height139-0272Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelvingLog in for availability1 * 1 Items - kr 1 150,00
Product Information
- Catalog #ForAvailabilityPrice
- Stationary post, SiteSelect™, stainless steel, 875 mm heightStationary post, SiteSelect™, stainless steel, 875 mm height139-0258Super Erecta Pro™ and Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelvingLog in for availability1 * 1 Items - kr 1 300,00
Product Information
- Catalog #ForAvailabilityPrice
- Stationary post, SiteSelect™, stainless steel, 875 mm heightStationary post, SiteSelect™, stainless steel, 875 mm height139-0257Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelvingLog in for availability1 * 1 Items - kr 1 150,00
Product Information
- Catalog #ForAvailabilityPrice
- Mobile post, SiteSelect™, chrome plated, 1385 mm heightMobile post, SiteSelect™, chrome plated, 1385 mm height139-0267Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelvingLog in for availability1 * 1 Items - kr 555,00
Product Information
- Catalog #ForAvailabilityPrice
- Mobile post, SiteSelect™, stainless steel, 857 mm heightMobile post, SiteSelect™, stainless steel, 857 mm height139-0273Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelvingLog in for availability1 * 1 Items - kr 1 300,00
Product Information
- Catalog #ForAvailabilityPrice
- Stationary post, SiteSelect™, chrome plated, 1385 mm heightStationary post, SiteSelect™, chrome plated, 1385 mm height139-0252Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, Super Erecta Pro™ shelvingLog in for availability1 * 1 Items - kr 555,00
Product Information
- Catalog #ForAvailabilityPrice
- Stationary post, SiteSelect™, stainless steel, 2200 mm heightStationary post, SiteSelect™, stainless steel, 2200 mm height139-0262Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelvingLog in for availability1 * 1 Items - kr 2 075,00
Product Information
- Catalog #ForAvailabilityPrice
- Mobile post, SiteSelect™, chrome plated, 191 mm heightMobile post, SiteSelect™, chrome plated, 191 mm height139-0263Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelvingLog in for availability1 * 1 Items - kr 345,00
Product Information
- Catalog #ForAvailabilityPrice
- Stationary post, SiteSelect™, chrome plated, 191 mm heightStationary post, SiteSelect™, chrome plated, 191 mm height139-0248Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelvingLog in for availability1 * 1 Items - kr 345,00
Product Information
- Catalog #ForAvailabilityPrice
- Mobile post, SiteSelect™, stainless steel, 1385 mm heightMobile post, SiteSelect™, stainless steel, 1385 mm height139-0274Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, Super Erecta Pro™ shelvingLog in for availability1 * 1 Items - kr 1 425,00
Product Information
- Catalog #ForAvailabilityPrice
- Mobile post, SiteSelect™, stainless steel, 370 mm heightMobile post, SiteSelect™, stainless steel, 370 mm height139-0271Super Erecta Pro™, Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® solid shelvingLog in for availability1 * 1 Items - kr 865,00
Product Information
- Catalog #ForAvailabilityPrice
- Mobile post, SiteSelect™, chrome plated, 370 mm heightMobile post, SiteSelect™, chrome plated, 370 mm height139-0264Super Erecta Pro™, Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® solid shelvingLog in for availability1 * 1 Items - kr 385,00
Product Information
- Catalog #ForAvailabilityPrice
- Stationary post, SiteSelect™, stainless steel, 370 mm heightStationary post, SiteSelect™, stainless steel, 370 mm height139-0256Super Erecta Pro™, Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® solid shelvingLog in for availability1 * 1 Items - kr 865,00
Product Information
- Catalog #ForAvailabilityPrice
- Stationary post, SiteSelect™, stainless steel, 1895 mm heightStationary post, SiteSelect™, stainless steel, 1895 mm height139-0261Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, Stationary shelving, Super Erecta Pro™ shelvingLog in for availability1 * 1 Items - kr 1 775,00
Product Information
- Catalog #AvailabilityPrice
- Post clamps, joins units together for maximum strength, zinc-platedPost clamps, joins units together for maximum strength, zinc-platedMETO9994ZLog in for availability1 * 1 Items - kr 170,00
Product Information
- Catalog #ForAvailabilityPrice
- Mobile post, SiteSelect™, metroseal 3, 1880 mm heightMobile post, SiteSelect™, metroseal 3, 1880 mm heightMETO74UPK3Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta Pro™ shelvingLog in for availability1 * 1 Items - kr 745,00
Product Information
- Catalog #ForAvailabilityPrice
- Mobile post, SiteSelect™, chrome plated, 699 mm heightMobile post, SiteSelect™, chrome plated, 699 mm height139-0265Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelvingLog in for availability1 * 1 Items - kr 465,00
Product Information
- Catalog #ForAvailabilityPrice
- Stationary post, SiteSelect™, stainless steel, 1385 mm heightStationary post, SiteSelect™, stainless steel, 1385 mm height139-0259Super Erecta Pro™ and Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelvingLog in for availability1 * 1 Items - kr 1 425,00
Product Information
- Catalog #ForAvailabilityPrice
- Stationary post, SiteSelect™, chrome plated, 699 mm heightStationary post, SiteSelect™, chrome plated, 699 mm height139-0250Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelvingLog in for availability1 * 1 Items - kr 465,00
Product Information
- Catalog #AvailabilityPrice
- Heavy duty aluminum split sleeves with zinc ringsHeavy duty aluminum split sleeves with zinc ringsMETO9986HZLog in for availability1 * 1 Items - kr 615,00