Semi-prep column chiralpak® if 250 x 20 mm 5 µm sfc 1 * 1 items
Catalog # DAIC86445
Supplier: CHIRAL TECHNOLOGIES
Semi-prep column chiralpak® if 250 x 20 mm 5 µm sfc 1 * 1 items
Catalog # DAIC86445
Supplier: CHIRAL TECHNOLOGIES
Frequently Bought Together
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Snap Cap Vials
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
(S,S) Whelk-O 1 100x4.6 3.5um 1 * 1 items
Supplier: HICHROM
Liquid nitrogen storage vessels, TP
Supplier: Air Liquide