Lead(II) nitrate ≥99%
Glass bottle for solids, 25 kg
Supplier: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Purity: 99% CAS No.: 10099-74-8 Molecular Formula: Pb(NO3)2 Formula Weight: 331.20 Odour: Odourless Melting Point: 470° UN No.: 1469 MDL No.: MFCD00011153
Synonymer: Lead dinitrate
- CAS Number:10099-74-8
- Formel:Pb(NO₃)₂
- MDL-nummer:MFCD00011153
- Molecular Weight:331.21 g/mol
- Merck-indeksen:14,5414
- Molten Pt:470 °C
- Tetthet:4.530
- Lagringstemperatur:Ambient
- EINECS:233-245-9
- U.N:1469
- ADR:5.1,II
Documents
Spesifikasjoner
Spesifikasjoner
- Pack type:Glass bottle for solids
- Pk:25 kg



