Order Entry
Norway
Orders LinkContactUsLinkComponent
4-Methylmorpholine ≥99.0% (by GC)

4-Methylmorpholine ≥99.0% (by GC)

Glass bottle, 500 mL
Catalog # TCIAM0370-500ML
Flammable
Corrosive
Irritant
Supplier:   TCI
4-Methylmorpholine ≥99.0% (by GC)
Catalog # TCIAM0370-500ML
Supplier:  TCI
CAS Number:  109-02-4
4-Methylmorpholine ≥99.0% (by GC)
4-Methylmorpholine ≥99.0% (by GC)

Glass bottle, 500 mL

CAS Number:  109-02-4
Price
kr 420,00
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  NMM, N-Methylmorpholine
  • CAS Number:109-02-4
  • Formula:C₅H₁₁NO
  • MDL Number:MFCD00006175
  • Molecular Weight:101.15 g/mol
  • Merck Index:14,06277
  • Boiling Pt:116°C
  • Melting Pt:–66 °C
  • Density:0,922 g/cm³ (20 °C)
  • Flash Pt:18°C
  • Storage Temperature:Ambient
  • EINECS:203-640-0
  • UN:2535
  • ADR:3,II
Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 99.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications