Fluorescein
Glass bottle, 25 g
Catalog # TCIAF0095-25G
Supplier: TCI
Jump to:
Product Details & Documents
Synonyms: 3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid
- CAS Number:2321-07-5
- Formula:C₂₀H₁₂O₅
- MDL Number:MFCD00005050
- Molecular Weight:332.31 g/mol
- Merck Index:14,04159
- Melting Pt:315°C
- Storage Temperature:Ambient
- EINECS:219-031-8
Documents
Specifications
Specification Test Results
- Appearance:Orange to Amber to Dark red powder to crystal
- Solubility in hot EtOH (95vol%):almost transparency
- Drying loss:max. 1.0 %
- Ignition residue (Sulfate):max. 2.0 %
- Sensitiveness:to pass test
Specifications
- Pack type:Glass bottle
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



