Skip to Header
Skip to Main Content
Skip to Footer
Avansert
Avansert
SDS
Sertifikat
Produsentdata
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Ph100 - 100ml/min hydrogen gas generator, gc uk plug 1 * 1 items
Catalog # 564-0097
Supplier:
PEAK SCIENTIFIC
Expand image
Ph100 - 100ml/min hydrogen gas generator, gc uk plug 1 * 1 items
Catalog # 564-0097
Supplier:
PEAK SCIENTIFIC