Order Entry
Norway
Orders LinkContactUsLinkComponent
 

Pipette tips, GripTips

GREEN CHOICE, standard, 12,5 µl, -, 35,56 mm, 5 inserts, 384 each
  613-3203
Pipette tips, GripTips
  613-3203
Pipette tips, GripTips
Pipette tips, GripTips

GREEN CHOICE, standard, 12,5 µl, -, 35,56 mm, 5 inserts, 384 each

 
kr 2 750,00
   
1 * 1.920 Items
 

GripTips are manufactured from virgin medical grade materials and designed for use with the VIAFLO pipettes. Their features perfectly match the tip fitting to form a complete system. Without hammering, the GripTips literally snap onto the pipette's tip fitting, which prevents tips from falling off.

  • Reduced tip attachment and ejection forces for comfortable, stress-free pipetting
  • Extended length design enables easy access into tubes and deep well blocks
  • Sterile tips are certified free of RNase, DNase, pyrogens, PCR inhibitors and endotoxins
  • GREEN CHOICE refill system: environmentally friendly refill system to reuse the current tip racks and thus reduce plastic waste by 60%

 : Available in bulk packs, tip racks with single inserts in 96 and 384 fold configuration or as GREEN CHOICE refills.

 
  • Packed:
    5 inserts, 384 each
  • Sterile:
    -
  • Volume:
    12,5 µl
  • Length:
    35,56 mm
  • Seq:
    07
  • Tip style:
    GREEN CHOICE, standard