Order Entry
Norway
Orders LinkContactUsLinkComponent
Kjeldahl tablets

Kjeldahl tablets

Wieninger 3,5 g tablet, 96,5% Na₂SO₄; 1,5% CuSO₄; 2% Se, 250 Tab.
Catalog # 1.17958.0250
Supplier:   Merck
Kjeldahl tablets
Catalog # 1.17958.0250
Supplier:  Merck
CAS Number:  
Kjeldahl tablets

Wieninger 3,5 g tablet, 96,5% Na₂SO₄; 1,5% CuSO₄; 2% Se, 250 Tab.

CAS Number:  
Price
kr 3 260,00
Log in for å se tilgjengelighet og deres pris
1 * 1 Each
Product Details & Documents

In addition to the traditional 5 g Kjeldahl tablets, which are added as catalysts to the digestion mixture, Merck Millipore can offer the 3,5 g tablet. This gives the flexibility needed to find the optimal setting for the catalyst/sulphuric acid ratio.

  • The 3,5 g tablets are suitable for microdigestion
  • Easy to use PE packaging concept
Spesifikasjoner

Spesifikasjoner

  • Pk:
    250 Tab.
  • Description:
    Wieninger 3,5 g tablet
  • Contents:
    96,5% Na₂SO₄; 1,5% CuSO₄; 2% Se

Spesifikasjoner

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.