Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Logg inn
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Colour stripes red,Concept worktable frame 1 * 1 items
Catalog # SOVE860851-00
Leverandør:
TRESTON
Expand image
Colour stripes red,Concept worktable frame 1 * 1 items
Catalog # SOVE860851-00
Leverandør:
TRESTON