0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Cover, petridish, material quartzglass, diameter 120mm, height 15mm. 1 * 1 items
Catalog # QUTW1174
Supplier:
QUARTZTECHNISCHE WERKSTATTEN
Expand image
Cover, petridish, material quartzglass, diameter 120mm, height 15mm. 1 * 1 items
Catalog # QUTW1174
Supplier:
QUARTZTECHNISCHE WERKSTATTEN