Order Entry
Norway
Orders LinkContactUsLinkComponent
Human TNF alpha ELISPOT Kit

Human TNF alpha ELISPOT Kit

Human TNF alpha ELISPOT kit
Catalog # ABCAAB62945-96
Supplier:   Abcam
Human TNF alpha ELISPOT Kit
Catalog # ABCAAB62945-96
Supplier:  Abcam
CAS Number:  
Human TNF alpha ELISPOT Kit
Human TNF alpha ELISPOT Kit

Human TNF alpha ELISPOT kit

CAS Number:  
Price
kr 6 225,00
Log in for availability
1 * 1 Each
Product Details & Documents

Human TNF alpha ELISPOT Kit is a ELISPOT Kit for the sensitive detection of Human TNF alpha production in a single cell suspension.

Specifications

Specifications

  • Description:
    Human TNF alpha ELISPOT kit
  • Cat. No.:
    ABCAAB62945-96
  • Sample type:
    Suspension cells
  • Form:
    Pre-coated
  • Species reactivity:
    Human
  • Storage temperature:
    4 °C
  • Processing time:
    Multiple steps
  • Regulatory status:
    RUO
  • Antigen symbol:
    TNF alpha

Specifications