Order Entry
Norway
ContactUsLinkComponent
Metal Packed GC Column (General Configuration), phase none, matrix 80/100 Porapak Q support, L x O.D. x I.D. 6.0 ft (1.8 m) x 1/8 in . x 2.1 mm 1 * 1 Each
Catalog # 12437.
Supplier:  Merck
Metal Packed GC Column (General Configuration), phase none, matrix 80/100 Porapak Q support, L x O.D. x I.D. 6.0 ft (1.8 m) x 1/8 in . x 2.1 mm 1 * 1 Each
Catalog # 12437.
Supplier:  Merck
Related Information