Order Entry
Norway
ContactUsLinkComponent
Coated GC Packing, phase 5% Carbowax 20M, matrix 60/80 Carbopack B support, bottle of 15 g 1 * 15 g
Catalog # 11766.
Supplier:  Merck
Coated GC Packing, phase 5% Carbowax 20M, matrix 60/80 Carbopack B support, bottle of 15 g 1 * 15 g
Catalog # 11766.
Supplier:  Merck