Order Entry
Norway
Orders LinkContactUsLinkComponent
 

Storage vials

Standard vial with round interior and square exterior, 3,00 ml, 17,3×26,4 mm, No, 23 mm
  SAVI200-003-20
 :   SAVILLEX
Storage vials
  SAVI200-003-20
 :  SAVILLEX
Storage vials
Storage vials

Standard vial with round interior and square exterior, 3,00 ml, 17,3×26,4 mm, No, 23 mm

 
kr 270,00
   
1 * 1 Items
 

PFA vials with round or conical interior.

  • Ideal for storage and digestion in geochemistry
  • Flat base for maximum heating efficiency
  • Square exterior vials with hex closures have higher pressure rating

 : Closures can be ordered separately. Modifications may be available on request.

 
  • Cap included:
    No
  • Cap Size:
    23 mm
  • Ø×H:
    17,3×26,4 mm
  • Capacity:
    3,00 ml
  • Description:
    Standard vial with round interior and square exterior
  • Seq:
    01