Order Entry
Norway
ContactUsLinkComponent
Refrigeration bath circulator steel, sc100-a10, -10 to +100 degree c, 240 w cooling capacity at 20c, 5 l bath volume, 2.0 kw heater, pump 17 l/min & 300 mbar 3 years warranty 230v/50hz 1 * 1 items
Catalog # HAAK152-5101
Supplier:  HAAKE
Refrigeration bath circulator steel, sc100-a10, -10 to +100 degree c, 240 w cooling capacity at 20c, 5 l bath volume, 2.0 kw heater, pump 17 l/min & 300 mbar 3 years warranty 230v/50hz 1 * 1 items
Catalog # HAAK152-5101
Supplier:  HAAKE