Pipette tips
VistaRak™, pyrogen-free, RNase/DNase certified, 1250 µl, +, 5 racks, 192 each
Catalog # VIST4060-3132
Supplier: VISTALAB TECHNOLOGIES
Jump to:
Product Details & Documents
These tips to maximise the exceptional performance and benefits of ovation and MLA pipettes. All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time.
Specifications
- Packed:5 racks, 192 each
- Sterile:+
- Volume:1250 µl
- Seq:13
- Tip style:VistaRak™, pyrogen-free, RNase/DNase certified