Order Entry
Norway
ContactUsLinkComponent
perforated insert standard, stainless steel, raw load capacity (evenly distributed load) (kg): 75 for underbench cabinets with width 1100 mm for model(s): ub90, ub30 1 * 1 items
Catalog # MSGA9973
Supplier:  asecos
perforated insert standard, stainless steel, raw load capacity (evenly distributed load) (kg): 75 for underbench cabinets with width 1100 mm for model(s): ub90, ub30 1 * 1 items
Catalog # MSGA9973
Supplier:  asecos