kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
BUFFER ANTIGEN RETRIEVAL 10X PH 10 1 * 200 mL
DIABK002-200
:
DIAGNOSTIC BIOSYSTEM
BUFFER ANTIGEN RETRIEVAL 10X PH 10 1 * 200 mL
DIABK002-200
:
DIAGNOSTIC BIOSYSTEM