Order Entry
Norway
ContactUsLinkComponent
Vial, capacity 2ml, target snap-it, 11mm crimp/snap, clear, glass, flat bottom, diameter 12mm, height 32mm, usable volume 1.5ml, res idual volume <170µl, not patched
Catalog # CRMAC4011-5
Supplier:  CHROMACOL
Vial, capacity 2ml, target snap-it, 11mm crimp/snap, clear, glass, flat bottom, diameter 12mm, height 32mm, usable volume 1.5ml, res idual volume <170µl, not patched
Catalog # CRMAC4011-5
Supplier:  CHROMACOL